Assay Details
| Gene Symbol: Cdipt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138899.2 | NP_620254.1 | 1 - 2 | 441 | 101 | |
| XM_006230208.1 | XP_006230270.1 | 1 - 2 | 615 | 101 | ||
| GenBank mRNA | BC070876.1 | - | 1 - 2 | 441 | 7096385 | 101 |
| D82928.1 | - | 1 - 2 | 189 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC070876.1, D82928.1, NM_138899.2, XM_006230208.1 transcripts.
Set Membership: |
|

Cdipt
Genomic Map