Assay Details
| Gene Symbol: Prkrir | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191630.1 | NP_001178559.1 | 4 - 5 | 421 | 115 | |
| XM_008759719.1 | XP_008757941.1 | 6 - 7 | 869 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkrir
Genomic Map