Assay Details
| Gene Symbol: Taf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134735.1 | NP_001128207.1 | 4 - 5 | 584 | 84 | |
| GenBank mRNA | BC168203.1 | - | 4 - 5 | 584 | 7115651 | 84 |
More Information
Updated Mapping Information
Set Membership: |
|

Taf10
Genomic Map