Assay Details
| Gene Symbol: Rabep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030585.1 | NP_085074.1 | 2 - 3 | 297 | 156 | |
| GenBank mRNA | BC089795.1 | - | 2 - 3 | 283 | 7375879 | 156 |
| U34932.1 | - | 2 - 3 | 297 | 156 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabep2
Genomic Map