Assay Details
| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230186.2 | XP_006230248.1 | 14 - 15 | 2304 | 96 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P292711 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P16309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59261 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16309 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P292711 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P292711 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P16309 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P292711 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P16309 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P292711 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16309 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10710719 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710722 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710721 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710726 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710724 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710733 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710720 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710728 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710730 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10710718 | - | - |
| Rat Genome 230 2.0 Array | 1381922_at | - | - |
| Rat Genome 230 2.0 Array | 1381922_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381922_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381922_at | - | - |
| Rat Expression Set 230 | 1381922_PM_at | - | - |
| Rat Expression Set 230 | 1381922_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364677 | - | - |

Slc5a11
Genomic Map