Assay Details
| Gene Symbol: Itgax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230382.2 | XP_006230444.1 | 10 - 11 | 1745 | 64 | |
| XM_574569.6 | XP_574569.3 | 14 - 15 | 2069 | 64 | ||
More Information
Set Membership: |
|

Itgax
Genomic Map