Assay Details
| Gene Symbol: Sec23ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134859.1 | NP_001128331.1 | 5 - 6 | 1331 | 68 | |
| XM_006230387.1 | XP_006230449.1 | 5 - 6 | 1265 | 68 | ||
| XM_006230388.1 | XP_006230450.1 | 4 - 5 | 832 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sec23ip
Genomic Map