Assay Details
| Gene Symbol: LOC100909464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750611.3 | XP_003750659.3 | 4 - 5 | 626 | 83 | |
| Gene Symbol: Ppp2r5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001062510.5 | XP_001062510.4 | 7 - 8 | 1492 | 83 | |
| XM_006244570.2 | XP_006244632.1 | 7 - 8 | 1495 | 83 | ||
| GenBank mRNA | BC099800.1 | - | 6 - 7 | 784 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100909464
Genomic Map