Assay Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191582.1 | NP_001178511.1 | 26 - 27 | 5189 | 61 | |
| XM_006244518.2 | XP_006244580.1 | 26 - 27 | 5513 | 61 | ||
| XM_008766864.1 | XP_008765086.1 | 26 - 27 | 5513 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cul9
Genomic Map