Assay Details
| Gene Symbol: Gpr116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139110.1 | NP_620810.1 | 5 - 6 | 667 | 160 | |
| XM_006244594.2 | XP_006244656.1 | 5 - 6 | 882 | 160 | ||
| XM_006244597.2 | XP_006244659.1 | 5 - 6 | 881 | 160 | ||
| XM_008766832.1 | XP_008765054.1 | 5 - 6 | 884 | 160 | ||
| XM_008766833.1 | XP_008765055.1 | 5 - 6 | 809 | 160 | ||
| XM_008766834.1 | XP_008765056.1 | 5 - 6 | 885 | 160 | ||
| XM_008766835.1 | XP_008765057.1 | 5 - 6 | 883 | 160 | ||
| GenBank mRNA | AB019120.1 | - | 5 - 6 | 667 | 160 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr116
Genomic Map