Assay Details
| Gene Symbol: Mrps18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198756.2 | NP_942051.1 | 3 - 4 | 258 | 144 | |
| GenBank mRNA | BC058507.1 | - | 3 - 4 | 262 | 6889538 | 144 |
| FQ215521.1 | - | 4 - 5 | 437 | 144 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrps18a
Genomic Map