Assay Details
| Gene Symbol: Polh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108204.1 | NP_001101674.1 | 8 - 9 | 1318 | 63 | |
| XM_006244537.1 | XP_006244599.1 | 6 - 7 | 746 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P995906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142678 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P995906 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P995906 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P995906 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P995906 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23351 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 4840 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10921758 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921752 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921754 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921757 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921755 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921756 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921759 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921761 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921760 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921753 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10921751 | - | - |
| Rat Genome 230 2.0 Array | 1393823_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393823_at | - | - |
| Affymetrix HT Rat Focus Array | 1393823_at | - | - |
| Affymetrix HT Rat Focus Array | 1393823_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900002130.rn.1 | TC0900000236.rn.1 | RTA TC |
| Rat Expression Set 230 | 1393823_at | - | - |
| Rat Expression Set 230 | 1393823_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10921751 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21072802 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369597 | - | - |

Polh
Genomic Map