Assay Details
| Gene Symbol: LOC102555622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244504.2 | XP_006244566.1 | 1 - 2 | 724 | 66 | |
| Gene Symbol: Foxp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108788.1 | NP_001102258.1 | 1 - 2 | 272 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108788.1 transcripts.
Set Membership: |
|

LOC102555622
Genomic Map