Assay Details
| Gene Symbol: Trem2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244424.2 | XP_006244486.1 | 3 - 4 | 721 | 104 | |
| XM_006244425.2 | XP_006244487.1 | 3 - 4 | 718 | 104 | ||
| GenBank mRNA | FQ229837.1 | - | 3 - 4 | 550 | 104 | |
More Information
Set Membership: |
|

Trem2
Genomic Map