Assay Details
| Gene Symbol: Taf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108197.1 | NP_001101667.1 | 1 - 2 | 58 | 81 | |
| XM_008766860.1 | XP_008765082.1 | 1 - 2 | 59 | 81 | ||
| XM_008766861.1 | XP_008765083.1 | 1 - 2 | 59 | 81 | ||
| XM_008766862.1 | XP_008765084.1 | 1 - 2 | 59 | 81 | ||
| GenBank mRNA | FQ211123.1 | - | 1 - 2 | 55 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001108197.1, XM_008766860.1, XM_008766861.1, XM_008766862.1 transcripts.
Set Membership: |
|

Taf8
Genomic Map