Assay Details
| Gene Symbol: Taf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108197.1 | NP_001101667.1 | 2 - 3 | 216 | 64 | |
| XM_008766860.1 | XP_008765082.1 | 2 - 3 | 217 | 64 | ||
| XM_008766861.1 | XP_008765083.1 | 2 - 3 | 217 | 64 | ||
| XM_008766862.1 | XP_008765084.1 | 2 - 3 | 217 | 64 | ||
| GenBank mRNA | FQ211123.1 | - | 2 - 3 | 213 | 64 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Taf8
Genomic Map