Assay Details
| Gene Symbol: LOC100911564 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766920.1 | XP_008765142.1 | 4 - 5 | 629 | 234 | |
| Gene Symbol: Gnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017084.1 | NP_058780.1 | 4 - 5 | 597 | 234 | |
| GenBank mRNA | X06150.1 | - | 4 - 5 | 597 | 234 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911564
Genomic Map