Assay Details
| Gene Symbol: Spata31a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008359.2 | NP_001008360.2 | 3 | 67 | ||
| GenBank mRNA | AF511042.2 | - | 3 | 67 | ||
| BC085936.1 | - | 3 | 7115545 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Spata31a5
Genomic Map