Assay Details
| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013192.2 | NP_037324.1 | 196 | 83 | ||
| GenBank mRNA | AB073753.1 | - | 196 | 83 | ||
| AB073754.1 | - | 196 | 83 | |||
| AB073755.1 | - | 196 | 83 | |||
| AB073756.1 | - | 196 | 83 | |||
| U21087.1 | - | 196 | 83 | |||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB073753.1,AB073754.1,AB073755.1,AB073756.1,NM_013192.2,U21087.1 transcripts.
Set Membership: |
|

Kcnj6
Genomic Map