Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012520.2 | NP_036652.1 | 8 - 9 | 1192 | 55 | |
| GenBank mRNA | BC081853.1 | - | 8 - 9 | 1089 | 7109889 | 55 |
| M11670.1 | - | 8 - 9 | 1143 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cat
Genomic Map