Assay Details
| Gene Symbol: Ap5s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134618.1 | NP_001128090.1 | 1 - 2 | 291 | 83 | |
| XM_006235038.2 | XP_006235100.1 | 1 - 2 | 310 | 83 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134618.1,XM_006235038.2 transcripts.
Set Membership: |
|

Ap5s1
Genomic Map