Assay Details
| Gene Symbol: Ndufaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106500.1 | NP_001099970.1 | 2 - 3 | 856 | 71 | |
| XM_006234737.1 | XP_006234799.1 | 2 - 3 | 736 | 71 | ||
| XM_006234738.2 | XP_006234800.1 | 3 - 4 | 733 | 71 | ||
| XM_006234739.2 | XP_006234801.1 | 2 - 3 | 724 | 71 | ||
| XM_006234740.2 | XP_006234802.1 | 2 - 3 | 980 | 71 | ||
| XM_006234741.2 | XP_006234803.1 | 2 - 3 | 880 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufaf1
Genomic Map