Assay Details
| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047857.3 | NP_001041322.2 | 4 - 5 | 756 | 114 | |
| XM_006234405.2 | XP_006234467.1 | 4 - 5 | 683 | 114 | ||
| GenBank mRNA | DQ480743.1 | - | 4 - 5 | 759 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2e3
Genomic Map