Assay Details
| Gene Symbol: Stk35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107773.1 | NP_001101243.1 | 1 - 2 | 1351 | 54 | |
| XM_006234975.2 | XP_006235037.1 | 1 - 2 | 1409 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk35
Genomic Map