Assay Details
| Gene Symbol: Atg4al1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126298.1 | NP_001119770.1 | 1 - 2 | 38 | 125 | |
| GenBank mRNA | BC160890.1 | - | 1 - 2 | 38 | 7442516 | 125 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC160890.1,NM_001126298.1 transcripts.
Set Membership: |
|

Atg4al1
Genomic Map