Assay Details
| Gene Symbol: Atg4al1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126298.1 | NP_001119770.1 | 581 | 140 | ||
| GenBank mRNA | BC160890.1 | - | 581 | 7442516 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

Atg4al1
Genomic Map