Assay Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106493.2 | NP_001099963.1 | 2 - 3 | 228 | 61 | |
| XM_006234639.2 | XP_006234701.1 | 2 - 3 | 2727 | 61 | ||
| XM_006234641.2 | XP_006234703.1 | 2 - 3 | 302 | 61 | ||
| GenBank mRNA | BC166742.1 | - | 2 - 3 | 228 | 7937842 | 61 |
More Information
Updated Mapping Information
Set Membership: |
|

Ehf
Genomic Map