Assay Details
| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107746.1 | NP_001101216.1 | 1 - 2 | 59 | 67 | |
| XM_006234507.2 | XP_006234569.1 | 1 - 2 | 51 | 67 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107746.1,XM_006234507.2 transcripts.
Set Membership: |
|

Kbtbd4
Genomic Map