Assay Details
| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107746.1 | NP_001101216.1 | 2 - 3 | 673 | 68 | |
| XM_006234507.2 | XP_006234569.1 | 2 - 3 | 665 | 68 | ||
| XM_006234508.1 | XP_006234570.1 | 2 - 3 | 755 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kbtbd4
Genomic Map