Assay Details
| Gene Symbol: Prnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006235058.2 | XP_006235120.1 | 1 - 2 | 104 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235058.2 transcripts.
Set Membership: |
|

Prnd
Genomic Map