Assay Details
| Gene Symbol: Cyct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012840.3 | NP_036972.1 | 406 | 94 | ||
| GenBank mRNA | M20623.1 | - | 1 - 2 | 236 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyct
Genomic Map