Assay Details
| Gene Symbol: Mga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234797.2 | XP_006234859.1 | 9 - 10 | 3589 | 86 | |
| XM_006234798.2 | XP_006234860.1 | 9 - 10 | 4124 | 86 | ||
| XM_006234799.2 | XP_006234861.1 | 9 - 10 | 3648 | 86 | ||
| XM_006234800.2 | XP_006234862.1 | 9 - 10 | 3589 | 86 | ||
| XM_008762120.1 | XP_008760342.1 | 9 - 10 | 3890 | 86 | ||
More Information
Set Membership: |
|

Mga
Genomic Map