Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199107.1 | NP_954538.1 | 13 - 14 | 1759 | 77 | |
| XM_006234562.2 | XP_006234624.1 | 12 - 13 | 3080 | 77 | ||
| XM_006234564.2 | XP_006234626.1 | 13 - 14 | 1819 | 77 | ||
| XM_006234566.2 | XP_006234628.1 | 13 - 14 | 1729 | 77 | ||
| XM_008762024.1 | XP_008760246.1 | 12 - 13 | 1892 | 77 | ||
| XM_008762025.1 | XP_008760247.1 | 12 - 13 | 2007 | 77 | ||
| GenBank mRNA | BC061762.1 | - | 13 - 14 | 1759 | 5621056 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map