Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199107.1 | NP_954538.1 | 5 - 6 | 499 | 69 | |
| XM_006234562.2 | XP_006234624.1 | 4 - 5 | 1820 | 69 | ||
| XM_006234564.2 | XP_006234626.1 | 5 - 6 | 559 | 69 | ||
| XM_006234566.2 | XP_006234628.1 | 5 - 6 | 559 | 69 | ||
| XM_008762024.1 | XP_008760246.1 | 4 - 5 | 632 | 69 | ||
| XM_008762025.1 | XP_008760247.1 | 4 - 5 | 747 | 69 | ||
| GenBank mRNA | BC061762.1 | - | 5 - 6 | 499 | 5621056 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map