Assay Details
| Gene Symbol: Tgm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108959.1 | NP_001102429.1 | 11 - 12 | 1860 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94365 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054074 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054074 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054074 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P368153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10839815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839813 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839822 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839820 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839823 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839818 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839817 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839819 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839821 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839824 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10839812 | - | - |
| Rat Genome 230 2.0 Array | 1378675_at | - | - |
| Rat Genome 230 2.0 Array | 1378675_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378675_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378675_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300012221.rn.1 | TC0300001706.rn.1 | RTA TC |
| Rat Expression Set 230 | 1378675_PM_at | - | - |
| Rat Expression Set 230 | 1378675_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353665 | - | - |

Tgm3
Genomic Map