Assay Details
| Gene Symbol: Katnbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109645.2 | NP_001103115.1 | 4 - 5 | 528 | 83 | |
| XM_006234712.2 | XP_006234774.1 | 4 - 5 | 581 | 83 | ||
| GenBank mRNA | BC161951.1 | - | 4 - 5 | 528 | 9026012 | 83 |
More Information
Updated Mapping Information
Set Membership: |
|

Katnbl1
Genomic Map