Assay Details
Gene Symbol: Lrrc4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107753.1 | NP_001101223.1 | 1295 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001107753.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
