Assay Details
| Gene Symbol: Zswim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011960.1 | NP_001011960.1 | 2 - 3 | 275 | 93 | |
| XM_006234435.2 | XP_006234497.1 | 2 - 3 | 309 | 93 | ||
| GenBank mRNA | BC079431.1 | - | 2 - 3 | 275 | 7114765 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|

Zswim2
Genomic Map