Assay Details
| Gene Symbol: Prnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012631.2 | NP_036763.1 | 1 - 2 | 49 | 95 | |
| XM_006235062.2 | XP_006235124.1 | 1 - 2 | 105 | 95 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_012631.2,XM_006235062.2 transcripts.
Set Membership: |
|

Prnp
Genomic Map