Assay Details
| Gene Symbol: Smox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134854.1 | NP_001128326.1 | 1 - 2 | 517 | 109 | |
| XM_006235063.2 | XP_006235125.1 | 2 - 3 | 409 | 109 | ||
| XM_006235064.2 | XP_006235126.1 | 2 - 3 | 408 | 109 | ||
| XM_006235065.2 | XP_006235127.1 | 2 - 3 | 408 | 109 | ||
| XM_008762165.1 | XP_008760387.1 | 2 - 3 | 425 | 109 | ||
| GenBank mRNA | BC160836.1 | - | 2 - 3 | 292 | 7456805 | 109 |
| BC161894.1 | - | 1 - 2 | 517 | 7193521 | 109 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Smox
Genomic Map