Assay Details
| Gene Symbol: Shf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234884.2 | XP_006234946.1 | 1 - 2 | 131 | 61 | |
| XM_006234885.2 | XP_006234947.1 | 1 - 2 | 131 | 61 | ||
| XM_008762216.1 | XP_008760438.1 | 2 - 3 | 434 | 61 | ||
| XM_008762217.1 | XP_008760439.1 | 1 - 2 | 131 | 61 | ||
| XM_008762218.1 | XP_008760440.1 | 2 - 3 | 434 | 61 | ||
More Information
Set Membership: |
|

Shf
Genomic Map