Assay Details
| Gene Symbol: Pcna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022381.3 | NP_071776.1 | 2 - 3 | 377 | 107 | |
| GenBank mRNA | BC060570.1 | - | 2 - 3 | 462 | 6891516 | 107 |
| FQ219845.1 | - | 2 - 3 | 458 | 107 | ||
| FQ219897.1 | - | 2 - 3 | 458 | 107 | ||
| FQ220101.1 | - | 2 - 3 | 458 | 107 | ||
| FQ220309.1 | - | 2 - 3 | 458 | 107 | ||
| FQ220835.1 | - | 2 - 3 | 458 | 107 | ||
| FQ220924.1 | - | 2 - 3 | 457 | 107 | ||
| FQ225620.1 | - | 1 - 2 | 82 | 107 | ||
| FQ226726.1 | - | 2 - 3 | 459 | 107 | ||
| FQ227500.1 | - | 2 - 3 | 458 | 107 | ||
| FQ227521.1 | - | 2 - 3 | 458 | 107 | ||
| FQ229469.1 | - | 2 - 3 | 458 | 107 | ||
| FQ229972.1 | - | 2 - 3 | 458 | 107 | ||
| FQ230190.1 | - | 2 - 3 | 458 | 107 | ||
| FQ232129.1 | - | 2 - 3 | 471 | 107 | ||
| FQ233125.1 | - | 2 - 3 | 458 | 107 | ||
| FQ233579.1 | - | 2 - 3 | 458 | 107 | ||
| FQ234183.1 | - | 2 - 3 | 459 | 107 | ||
| FQ234500.1 | - | 2 - 3 | 458 | 107 | ||
| FQ234529.1 | - | 2 - 3 | 458 | 107 | ||
| FQ234547.1 | - | 2 - 3 | 458 | 107 | ||
| FQ234986.1 | - | 2 - 3 | 458 | 107 | ||
| FQ235296.1 | - | 2 - 3 | 458 | 107 | ||
| Y00047.1 | - | 2 - 3 | 377 | 107 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcna
Genomic Map