Assay Details
| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111294.1 | NP_001104764.1 | 2 - 3 | 271 | 103 | |
| NM_001111295.1 | NP_001104765.1 | 2 - 3 | 205 | 103 | ||
| NM_198132.3 | NP_937765.1 | 2 - 3 | 271 | 103 | ||
| XM_008761911.1 | XP_008760133.1 | 2 - 3 | 220 | 103 | ||
| XM_008761912.1 | XP_008760134.1 | 2 - 3 | 220 | 103 | ||
| XM_008761913.1 | XP_008760135.1 | 2 - 3 | 266 | 103 | ||
| GenBank mRNA | AY363226.1 | - | 2 - 3 | 217 | 103 | |
| AY363227.1 | - | 2 - 3 | 151 | 103 | ||
| BC081878.1 | - | 2 - 3 | 211 | 7108128 | 103 | |
| CB725005.1 | - | 2 - 3 | 143 | 103 | ||
| FQ214617.1 | - | 2 - 3 | 167 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpa3
Genomic Map