Assay Details
| Gene Symbol: Hnrnpa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111294.1 | NP_001104764.1 | 1 - 2 | 157 | 110 | |
| NM_198132.3 | NP_937765.1 | 1 - 2 | 157 | 110 | ||
| XM_008761913.1 | XP_008760135.1 | 1 - 2 | 152 | 110 | ||
| GenBank mRNA | AY363226.1 | - | 1 - 2 | 103 | 110 | |
| BC081878.1 | - | 1 - 2 | 97 | 7108128 | 110 | |
| CB585714.1 | - | 1 - 2 | 169 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY363226.1, BC081878.1, NM_001111294.1, NM_198132.3, XM_008761913.1 transcripts.
Set Membership: |
|

Hnrnpa3
Genomic Map