Assay Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234419.2 | XP_006234481.1 | 1 - 2 | 2139 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006234419.2 transcripts.
Set Membership: |
|

Zfp385b
Genomic Map