Assay Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107736.1 | NP_001101206.1 | 2 - 3 | 1497 | 68 | |
| XM_006234416.2 | XP_006234478.1 | 2 - 3 | 1117 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp385b
Genomic Map