Assay Details
| Gene Symbol: Zfp385b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107736.1 | NP_001101206.1 | 6 - 7 | 2197 | 109 | |
| XM_006234414.2 | XP_006234476.1 | 7 - 8 | 1856 | 109 | ||
| XM_006234416.2 | XP_006234478.1 | 6 - 7 | 1817 | 109 | ||
| XM_006234419.2 | XP_006234481.1 | 7 - 8 | 3013 | 109 | ||
| XM_006234420.2 | XP_006234482.1 | 7 - 8 | 1382 | 109 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zfp385b
Genomic Map