Assay Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013203.1 | NP_001013221.1 | 1 - 2 | 673 | 72 | |
| GenBank mRNA | AY551091.1 | - | 1 - 2 | 673 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim44
Genomic Map