Assay Details
| Gene Symbol: Clcnkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239320.2 | XP_006239382.1 | 6 - 7 | 3248 | 73 | |
| XM_008764293.1 | XP_008762515.1 | 5 - 6 | 2916 | 73 | ||
| XM_008764294.1 | XP_008762516.1 | 5 - 6 | 1182 | 73 | ||
| XM_008764295.1 | XP_008762517.1 | 4 - 5 | 1327 | 73 | ||
| XM_008764296.1 | XP_008762518.1 | 4 - 5 | 1316 | 73 | ||
| GenBank mRNA | BC081698.1 | - | 3 - 4 | 871 | 73 | |
More Information
Set Membership: |
|

Clcnkb
Genomic Map