Assay Details
| Gene Symbol: Tmem54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034151.1 | NP_001029323.1 | 1 - 2 | 141 | 60 | |
| GenBank mRNA | BC101399.1 | - | 1 - 2 | 141 | 7933358 | 60 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC101399.1,NM_001034151.1 transcripts.
Set Membership: |
|

Tmem54
Genomic Map